SHEL LAB® SMI Series Microbiological Incubators, Sheldon Manufacturing
Supplier: Sheldon Manufacturing
SLM722 SLM722-E SLM1122 SLM1122-E SLM222 SLM222-E SLM622 SLM622-E SLM1222 SLM1222-E
76311-250EA 9012.08 CAD
76311-250 76311-252 76311-254 76311-256 76311-260 76311-262 76311-264 76311-266 76311-298 76311-300
SHEL LAB® SMI Series Microbiological Incubators, Sheldon Manufacturing
Incubators Standard Incubators
Microbiological incubator series has been updated for optimum performance. Ranging in sizes from 57 to 331 liters (2 to 11.7 cu.ft.), the SMI series is guaranteed to fit any workspace. Ideal for clinical, analytical, and general laboratory applications.
  • USB data logging capabilities
  • Independent over temperature set point and operational control override for additional safety
  • Side-mounted access port, 38 mm diameter (1.5")
  • Stainless steel interior construction for long life operation and easy cleaning
  • Condensed connection panel includes; 4 to 20 mA analog
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable