BioClean™ Legion Latex Gloves, Ansell
Supplier: Ansell Canada
BLA301-S BLA302-M BLA303-L BLA304-XL
CA75834-008CS 1539.6 CAD
CA75834-008 CA75834-010 CA75834-012 CA75834-014
BioClean™ Legion Latex Gloves, Ansell
Gloves
Offering comfort, flexibility and elbow length protection, these 400 mm gloves are the natural choice. With a textured surface for enhanced grip.
  • Textured fingers and palms
  • Easy double-donning
  • Non-particulating EasyTear packaging
Certifications: ISO 9001: 2008 facility certifications. Conforms to Category 3.Processed in an ISO Class 5 environment. Complies with EN 420:2003, EN 374-1, 2 & 3:2003, and EN 455-2:2009 glove standards. Suitable for contact with dry, alcoholic and acidic foodstuffs. CE 0120 marked.
Order Now
SPECIFICATIONS
Model BLA3
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable
Latex/Latex-free Latex
Powdered/Powder-free Powder-free
Material Natural rubber latex
Finish Textured Fingers and Palms
Cuff style Beaded
Finger thickness 0.20 mm
Palm thickness 0.17 mm
Sterility Non-Sterile
Food contact approved +