2-Methoxyestradiol ELISA Kit, Cayman Chemical Company

Supplier: Cayman Chemical Company

582261-480 582261-96
75817-382EA 3718.01 CAD
75817-382 75817-384
2-Methoxyestradiol ELISA Kit, Cayman Chemical Company
Assays ELISAs

A competitive assay that can be used for quantification of 2-methoxyestradiol in plasma, urine, and other sample matrices.


  • Detect 2-ME2
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure 2-ME2 levels down to 40 pg/ml
  • Colorimetric read at 405 to 420 nm


2-Methoxyestradiol (2-ME2) is a natural metabolite of estradiol produced by CYP450-dependant hydroxylation followed by catechol-O-methyltransferase-catalyzed O-methylation. Deficiency in catechol-O-methyltransferase is associated with pre-eclampsia. Therefore, 2-ME2 may be utilized as a plasma and urinary diagnostic marker for this disease, as well as a therapeutic supplement for its prevention. Several clinical trials have demonstrated the antitumor effects of 2-ME2 in prostate cancer, breast cancer, renal cell carcinoma, ovarian cancer, glioblastoma, and multiple myeloma. Other studies indicate that 2-ME2 exhibits more potent cardioprotective effects than estradiol. Cayman’s 2-Methoxyestradiol ELISA Kit is a competitive assay that can be used for quantification of 2-ME2 in plasma, urine, and other sample matrices.


Delivery information: Kit contains antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein 2-Methoxyestradiol
Primary antibody reactivity
Assay Type Competitive
Host
Sensitivity 40 pg/ml
Detection range 13.1 - 8000 pg/ml
Mid point 330 - 390 pg/ml
Developmental time 90 - 120 min
Incubation time 18 hr

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR