High Viscosity Standards, Cannon Instruments
Supplier: CANNON INSTRUMENT COMPANY CA
9727-E25.016
9727-E25.032
9727-E25.128
9727-E25.500
9727-E42
CA10021-358EA
184.78
CAD
CA10021-358
CA10021-360
CA10021-362
CA10021-364
CA47735-156
High Viscosity Standards, Cannon Instruments
Standards
Viscometry Standards
These certified standards are useful for calibration and verification procedures.
- For asphalt and polymer applications
Certifications: Meets the specifications of ASTM D2170 and ASTM D2171.