Anti-TGF-beta RII Mouse Monoclonal Antibody [clone: 176510]

Supplier: Novus Biologicals
MAB2411-SP MAB2411-500
103296-126EA 338.46 CAD
103296-126 103351-272
Anti-TGF-beta RII Mouse Monoclonal Antibody [clone: 176510]
Antibodies
TGF-beta RII is a membrane-bound serine/threonine kinase. Upon ligand binding, TGF-beta RII interacts with TGF-beta RI to form the heteromeric signaling complex that transduces TGF-beta signals. A splice variant of the type II receptor, TGF-beta RIIb, containing a 25 amino acid residue insertion near the N-terminus of the mature protein has also been described.

  • Reconstitute at 0.5 mg/ml in sterile PBS

Detects human TGF-beta RII and TGF-beta RIIb in direct ELISAs and Western blots. In direct ELISAs and western blots, this antibody does not cross-react with recombinant human TGF-beta RIII, recombinant mouse (rm) TGF-beta RII, or rmTGF-beta RI.

Caution: For research use only and is not for use in humans or in clinical diagnosis.

Type: Primary
Antigen: TGFBR2
Clonality: Monoclonal
Clone: 176510
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR