Pioneer® Semi Micro PX Balances, Ohaus
Supplier: Ohaus
30475733 30475734
76289-360EA 10078.88 CAD
76289-360 76289-362
Pioneer® Semi Micro PX Balances, Ohaus
Balances and Scales
The Pioneer PX5 combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
  • The PX5 is durably constructed for versatile, long-term use
  • Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan
  • To capacity by subtraction (tare range)
  • Internal calibration
The PX5 is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
The PX5 features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity for easy communication.
Certifications: IEC/EN 61010-1; CAN/CSA C22.2 61010-1; UL 61010-1.
Ordering information: Manufacturer’s two-year parts and labor warranty (U.S. and Canada). Accessories not included.