HyClone™ Newborn Bovine Calf Serum, New Zealand, HyClone products (Cytiva)
Supplier: Cytiva
SH30401.02
CA76236-350EA 225.66 CAD
CA76236-350 CA76236-342 CA76236-344 CA76236-348
HyClone™ Newborn Bovine Calf Serum, New Zealand, HyClone products (Cytiva)
Sera (Serum)
Newborn bovine calf serum is a cost-effective alternative to foetal bovine serum (FBS). The serum is sourced from animals that are typically less than 10 days old.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature Less than –10 °C