BioClean™ Excell Sterile Nitrile Cleanroom Gloves, Ansell
Supplier: Ansell Canada
BEXS-100 BEXS-60 BEXS-65 BEXS-70 BEXS-75 BEXS-80 BEXS-85 BEXS-90
CA75972-596CS 1382.19 CAD
CA75972-596 CA75972-598 CA75972-600 CA75972-602 CA75972-604 CA75972-606 CA75972-608 CA75972-610
BioClean™ Excell Sterile Nitrile Cleanroom Gloves, Ansell
Gloves
Gloves are resistant to a range of chemicals and anti-static. Hand specific to reduce wearer hand fatigue, the gloves feature a textured surface enabling good grip and dexterity.
  • Accelerator-free
  • Non-particulating EasyTear packaging
  • External glove surface has textured fingers and palms
  • Internal glove surface is chlorinated
Certifications: ISO 9001: 2008 facility certifications. Conforms to Category 3. Processed in an ISO CLASS 4 and EU GMP GRADE A. Complies with EN 420:2003, EN 374-1, 2 & 3:2003, and EN 455-2:2009 glove standards. Sterilized in accordance with ISO 11137-1:2006. Suitable for contact with all foodstuffs. CE 0120 marked. ESD properties