BucketBinder Multi-Bucket Systems, Micronova
Supplier: Micronova
BB-DBL-DGRE BB-DBL-DBYE BB-TR-DBREYE
76289-874EA 1290.02 CAD
76289-874 76289-876 76289-878
BucketBinder Multi-Bucket Systems, Micronova
Mops
A cost efficient, ergonomic, and simple multi-bucket system that replaces the need for bulky carts.
  • Easy clip action allows attachment before or after bucket is filled with disinfectant
  • Polyketone hooks firmly secure two or three rectangular buckets together
  • Stainless steel base plate attaches at caster for support
  • Sturdy enough to secure two or three buckets and easy to maneuver with up to 15 liters of liquid in each bucket
  • Lightweight; less than 1 kg per pair
  • Allows scale up or down as required
  • Autoclavable
BucketBinder kits include two or three polypropylene buckets with casters (22 l capacity), and one or two sets of BucketBinder bucket connectors. The system offers easy movement, cleaning, and storage. Works well in both small and large controlled environments and the sterile core.
Will house W-94 flat mop Megawringer™, W-9, W-92 flat wringers or W-9B BoxWringer for string mops. Complements all the Micronova M-Zone FlatMops™, SnapMops™ and SlimLine™ mops.
Accessories information: B-7 polypropylene buckets with autoclavable casters available in five colors, blue, green, red, green and gray are available as replacement buckets.