BioClean™ Emerald Sterile Nitrile Cleanroom Gloves, Ansell
Supplier: Ansell Canada
BENS6.0 BENS6.5 BENS7.0 BENS7.5 BENS8.0 BENS8.5 BENS9.0 BENS10.0 BENS6.0 BENS6.5 BENS7.0 BENS7.5 BENS8.0 BENS8.5 BENS9.0 BENS10.0
75972-582CS 1286.1 CAD
75972-582 75972-584 75972-586 75972-588 75972-590 75972-592 75972-594 75972-648 CA75972-582 CA75972-584 CA75972-586 CA75972-588 CA75972-590 CA75972-592 CA75972-594 CA75972-648
BioClean™ Emerald Sterile Nitrile Cleanroom Gloves, Ansell
Gloves
Gloves are resistant to a range of chemicals, can be easily double-donned, and are accelerator-free to reduce the risk of latex allergies. The hand specific shape ensures wearer comfort and reduces hand fatigue, while enabling good dexterity.
  • Powder, sulfur and accelerator free
  • Textured finish with a smooth feel
  • Anti-static, no natural latex proteins
  • Internal glove surface is powder-free and chlorinated
Sterilized by gamma radiation, easy double-donning.
Certifications: ISO 9001: 2008 facility certifications. Conforms to Category 3. Processed in an ISO Class 4 environment. Complies with EN 420:2003, EN 374-1, 2 & 3:2003, and EN 455-2:2009 glove standards. Permeation standard ASTM D6978-05 for cytotoxic drugs. Sterilized in accordance with ISO 11137-1:2006. Suitable for contact with all foodstuffs. CE 0120 marked. EU GMP Grade A compatible.