pH Buffers, YSI
Supplier: XYLEM/YSI
3821
003822
03823
23202-821EA
41.08
CAD
23202-821
23202-822
23202-823
pH Buffers, YSI
Buffers
pH Calibration Buffers
pH buffers for pH probe calibration.
Each buffer case contains 6 individual pints (plastic). Buffer cases are available with a single buffer type or with an assortment of buffers (2 pints of pH 4, 7, and 10 buffers).