HiScale Columns Packed with Superdex Increase or Superose Increase Resins, Cytiva
Supplier: Cytiva
29321904 29321905 29321906 29321903 29321907 29321908
CA76308-948EA 28206.56 CAD
CA76308-948 CA76308-950 CA76308-952 CA76308-974 CA76308-976 CA76308-978
HiScale Columns Packed with Superdex Increase or Superose Increase Resins, Cytiva
Chromatography Columns
These HiScale columns packed with Superdex Increase or Superose Increase resins are used to purify larger volumes of up to 8.5 ml.
  • High-resolution SEC columns: Small particle size and narrow particle size distribution of the packed resin ensures highly pure protein
  • HiScale columns packed with SEC resins on demand, please request a quote
HiScale columns are packed on demand for you with Superdex 200 Increase, Superdex 75 Increase, and Superose 6 Increase SEC (gel filtration) resins. The columns are available in two sizes: 16 × 400 mm and 26 × 400 mm. These two column sizes enable purification of sample volumes up to 3.2 and 8.5 ml, respectively.