CellXpert® C170 Cell Culture CO₂ Incubators

Supplier: Eppendorf
CellXpert®
2231000759 6734010045 6734010045 2231000759 6734010015 6734010015
CA77321-656EA 16143.17 CAD
77321-656 CA77321-656 76308-822 MPCA76308-823 76460-802 CA76308-822
CellXpert® C170 Cell Culture CO₂ Incubators
Incubators CO₂ Incubators
No room for contamination. The CellXpert C170 CO₂ incubator has a seamless chamber with no fan and an easy-to-clean interior with 140 °C High-temperature disinfection. Grow your cells consistently with fast recovery and temperature uniformity for more reproducible results.

  • Easy cleaning: Seamless chamber, easy disassembly in 40 seconds
  • Effective contamination protection: 140 °C High temperature disinfection (HTD), no internal fan-associated HEPA-filters necessary
  • Temperature uniformity: Innovative heating concept with multiple independent temperature sensors - verified at 27 spots inside the incubator according to German DIN 12880 norm
  • Fast recovery after door opening: Temperature and CO₂ in less than five minutes - no regulation overshoot for sample safety
  • Enhanced vibration and turbulence protection: No fan inside
  • Save money: Up to 25% more usable space by fanless design, no internal expendable parts (e.g. fan-associated HEPA-filters), significantly reduced gas consumption

Today's cell culture requires more than technical solutions to keep operational costs under control, optimize cell growth conditions, and efficiently prevent contamination.

The CellXpert® CO₂ incubator family was designed with the needs of modern cell culture laboratories in mind - to reduce time-to-market or publication with minimal efforts and resources. They are the perfect partner for demanding applications like assay development or culture of stem cells, primary cells, or 3D organoids and tissues.

If more options are needed (e.g. O₂ control, humidity monitoring, copper interior), 180 °C disinfection, or future flexibility - consider the CellXpert® C170i.

Chamber material: Deep-drawn (seamless) stainless steel
CO₂ uniformity: ±0.1%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR