Test Tube Racks for Open Air, Incubating and Incubating Cooling Shakers, OHAUS®
Supplier: OHAUS CORP.
30400110 30400111 30400112 30400113 30400105 30400106 30400107 30400108 30400109
470235-344EA 726 CAD
470235-344 470235-346 470235-348 470235-350 470235-352 470235-354 470235-356 470235-358 470235-360
Test Tube Racks for Open Air, Incubating and Incubating Cooling Shakers, OHAUS®
Racks Tube Racks
Test tube racks can be used for open air, incubating and incubating cooling shakers.
  • Pivoting rack has adjustable angle, custom tilt
  • Stainless steel holder includes removable plastic rack
  • Rack is easily removed to transport from work area to shaker
Full size stationary racks are constructed of PVC coated steel and it also includes hardware for easy attachment to platforms.