Micro Funnel, Hach®
Supplier: Hach
25843-35
CA25843-35EA 14.14 CAD
CA25843-35
Micro Funnel, Hach®
Funnels Filtration Funnels Micro Filter Funnels
Ideal for micro work, filling burets, manometers, and small bottles.
  • Polypropylene construction
  • Suitable for micro work
  • Steam autoclavable at 121 °C (250 °F)