KIMAX® KimCote® Squibb Separatory Funnels with Glass Stopper, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
KC29048T-125 KC29048T-250 KC29048T-500 KC29048T-1000 KC29048T-2000
89097-430CS 2183.49 CAD
89097-430 89097-432 89097-434 89097-436 89097-438
KIMAX® KimCote® Squibb Separatory Funnels with Glass Stopper, DWK Life Sciences
Funnels Separating Funnels
KIMAX® KimCote® plastic-coated, pear-shaped funnel.
Lower stems have an I-Ø large enough so that a column of liquid will 'break' with the stopcock closed, thereby giving a more complete separation, and eliminating the necessity of emptying the funnel.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: Meets ASTM Specification E1096, Type IV requirements.
Delivery information: Supplied with a [ST] ground glass stopper.