Ungraduated Addition Funnels with Pressure Equalizing Line, DWK Life Sciences
Supplier: DWK Life Sciences
634000-0125 634000-0250 634000-0500 634000-1000 634000-0060
KT634000-0125EA 673.91 CAD
KT634000-0125 KT634000-0250 KT634000-1000 KT634000-0060 KT634000-0500
Ungraduated Addition Funnels with Pressure Equalizing Line, DWK Life Sciences
Funnels Dropping Funnels
Cylindrical shaped funnels.
  • Glass stopcock
  • Standard taper upper joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirement.