Ribbed, Standard Stem Funnels, Chemglass
Supplier: Chemglass
CG-1770-01 CG-1770-02 CG-1770-03 CG-1770-04
89028-794EA 218.89 CAD
89028-794 89028-796 89028-798 89028-800
Ribbed, Standard Stem Funnels, Chemglass
Funnels Filtration Funnels Filter Funnels
Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock.
  • Parallel stem measures 8 mm O-Ø
  • Steam autoclavable at 121 °C (250 °F)