Conductive Liquid Handling (LiHa) Disposable Tips, Tecan
Supplier: TECAN US INC. CA
30000630 30057816 10612554 30000631 30057817 10612555 30057823 30057822 10612552 30057814 10612553 30000629 30057815 30057821 30057820 30000627 30083400 30083401 30032115 30057811 30032114 30057813 30057819 30057818 30059897 30065423 30032115 30057813 30057815
CA10024-072CS 1205.94 CAD
CA10024-072 CA10024-074 CA10024-076 CA10024-078 CA10024-080 CA10024-082 CA10024-148 CA10024-152 CA10024-158 CA10024-160 CA10024-164 CA10024-166 CA10024-168 CA10024-170 CA10024-172 CA10024-174 CA10024-176 CA10024-178 CA10024-180 CA10024-182 CA10024-184 CA10024-186 CA10024-188 CA10024-190 CA10024-192 CA10024-194 MSPP-30032115 MSPP-30057813 MSPP-30057815
Conductive Liquid Handling (LiHa) Disposable Tips, Tecan
Pipette Tips
These disposable tips are carefully designed, produced and validated to ensure reliable performance using Liquid Handling (LiHa) Arms and Flexible Channel (FCA) arms on Tecan platforms.
  • Available in a variety of tip volumes, with or without filters
  • Made of conductive polypropylene
  • Allow capacitive liquid level detection
  • Full product traceability
Tecan standard purity tips are supplied in space-saving cardboard packages and are used in applications such as ELISA, sample handling and biochemical screening.
Tecan pure purity tips are packaged in blister packs, and each lot of tips is tested and certified free from human DNA, DNase, RNase and PCR inhibitors by an external accredited laboratory and are used in applications such as genomics research, forensics and molecular diagnostics.
Tecan sterile purity tips are tested on a lot-by-lot basis in an external accredited laboratory, and are free from human DNA, DNase, RNase and endotoxins. These are certified to be sterile, with a guaranteed Sterility Assurance Level (SAL) of 10⁻⁶ and are supplied packaged in ANSI/SLAS-format boxes and are used in applications such as cell biology, microbiology and cell-based screening.
Delivery information: Each tray of Tecan disposable tips is supplied with a Certificate of Conformity (CoC).