arium® mini Essential Ultrapure Water Systems, Sartorius
Supplier: Sartorius
H2OMUSETUS H2OMUUVTUS H2OMUUVSETUS H2OMUTUS
76307-492EA 8921.28 CAD
76307-492 76307-494 76307-496 76307-662
arium® mini Essential Ultrapure Water Systems, Sartorius
Water Purification Systems Pure and Ultra Pure Water Systems
The arium® mini Essential Ultrapure Water benchtop system is designed for direct connection to a pre-treated water supply.
  • Large touch display with self-explanatory navigation
  • Direct access to all available dispensing options (direct/volume/favorite)
  • Produces ASTM Type 1 ultrapure (18.2 MOhm) water
  • Small footprint, only 28 cm width
  • Volume dispensing from 50 ml
  • Delivers a flow rate of one liter per minute
The arium® mini Essential system is directly connected to a pre-treated (DI/RO/EDI) centralized or decentralized feed water supply. Due to the slim design (28 cm width), the system saves space and can be installed almost anywhere in the lab. The system is equipped with a high-resolution, touch-activated color display. Easy-to-understand icons guide the user intuitively through the menu. A favorites function automatically stores the volume last dispensed. This increases efficiency and prevents errors during repeated dispensing of identical volumes. Optional built-in UV lamp and individually usable ultrafilter. The system is suitable for general-purpose and analytical applications, and is ideal for daily requirements involving less than 10 liters.
Methods: Adsorption by Spherical Activated Carbon, Catalyst, Reverse Osmosis, Ion Exchange, Optional Uv Irradiation and by Point of Use Particle-Removing Filtration/Sterile Filtration
Delivery information: All accessories are included with purchases of H2O-MU-SET-US and H2O-MU-UV-SET-US only. Set accessories include Scientific Pack, 5 l bag, and sterile final filter.