VWR® Conical Bottom Centrifuge Tubes, PP
Supplier: VWR International
76234-744 76234-746 76234-748
76234-744CS 183.18 CAD
76234-744 76234-746 76234-748
VWR® Conical Bottom Centrifuge Tubes, PP
Tubes Centrifuge Tubes
VWR® Conical bottom centrifuge tubes are made from high-grade gamma resistant polypropylene, in compliance with USP Class VI regulations.
  • Easy to read engraved graduations
  • Max RCF up to 7500×g
  • Autoclavable at 121 °C and freezable to −196 °C
  • Leak-proof and latex-free
  • Three years shelf life
  • Manufactured in a class 100000 cleanroom environment
Centrifuge tubes are available in 225, 250 and 500 ml sizes. The 225 and 250 ml sizes feature a plug seal cap. The 500 ml size features a flat seal cap. All caps are made from polyethylene. Sterilized by gamma irradiation to SAL 10⁻⁶ (ISO 11137).
Certifications: Meets the requirements of BSE/TSE. Certified DNase/RNase free and non-pyrogenic.
Caution: Height of 250 ml tubes has been changed from November 6th 2022, lot number: 221106334A.