Erlenmeyer Shaker Flasks, Greiner Bio-One
Supplier: Greiner Bio-One
679501 679502 679503 679504 679505 679506 679511 679512 679513 679514 679515 679516
76266-256CS 363.32 CAD
76266-256 76266-258 76266-260 76266-262 76266-306 76266-308 76266-310 76266-312 76266-314 76266-316 76266-318 76266-320
Erlenmeyer Shaker Flasks, Greiner Bio-One
Flasks Erlenmeyer Flasks
These erlenmeyer shaker flasks are ideal for suspension culture of mammalian cells, plants, microbes and microorganisms. The shaker flasks feature a patented 2-in-1 DuoCAP® which allows for flexibility between vented and non-vented applications.
  • Individually packaged and sterile to SAL 10-6 for long term culture applications
  • Molded-in graduations for easy volumetric reference
  • Non-pyrogenic, non-cytotoxic, and nuclease-free
Each flask is manufactured utilizing an optically clear, inert polycarbonate resin which can be disposed of after single use or be made reusable when autoclaved.
Ordering information: Available with flat or baffled base for increased agitation and aeration.