QuickPicker Core Sampler, Bürkle
Supplier: BURKLE INC
5322-1300 5322-0300 0318-0250 0319-0250
76311-032EA 774.47 CAD
76311-032 76311-034 76311-036 76311-038
QuickPicker Core Sampler, Bürkle
Samplers Solid Samplers
The QuickPicker core sampler is particularly suitable in the event that bulk goods are taken directly from sacks or bags, and have to be filled into the original sampling bottle directly. It is uncomplicated in its use and cleaning. No undercuts or edges mean no sample residues, which could falsify subsequent samples. It is ideal for quality control.
  • Insertion depth is 30 cm (11.81"), with a 25 mm (0.98") diameter
  • Removable tip for easy cleaning
  • Accessory sampling bottles are available in PE or PP formats
The QuickPicker is available in eitherMore Product Information