Containers, CTH Series, Eagle Stainless
Supplier: Eagle Stainless Container
CTH-24-316L-J CTH-30-316L-J CTH-36-316L-J CTH-43-316L-J CTH-47-316L-J CTH-47H-316L-J CTH-565H-316L-J CTH-565-316L-J
13308-240EA 1204.86 CAD
13308-240 13308-242 13308-244 13308-246 13308-248 13308-250 13308-280 13308-284
Containers, CTH Series, Eagle Stainless
Sample Containers General Purpose Containers
Containers, CTH series with clip down type closure.
  • Made of 316L stainless steel container and with lid
  • Electropolished finish (10 Ra)
  • With passivation option
Seamless sanitary interior and exterior with radiused bottom-corners and has 304 stainless steel clips with hollow side handles.
Air/water-tight seal with platinum-cured silicone gasket (Class VI Certification CTH-18 through CTH-47H).
Order Now
SPECIFICATIONS
Environmentally Preferable