Caps, GL Thread with Aperature, Chemglass
Supplier: Chemglass
CG-195-01 CG-195-02 CG-195-03 CG-195-04 CG-195-05
80061-836EA 6.74 CAD
80061-836 80061-838 80061-840 80061-842 80061-844
Caps, GL Thread with Aperature, Chemglass
Closures Bottle Closures
Cap is made from PBT (thermoplastic polyester). It can be combined with the sealing ring will accommodate tubing of the listed diameter.
  • Open top cap for use with GL screw threads
  • Can withstand temperatures from –45 to 200 °C