tetranor-PGDM ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical Company
501001-480 501001-96
75816-826EA 3237.64 CAD
75816-826 75816-828
tetranor-PGDM ELISA Kit, Cayman Chemical
Assays ELISAs
A competitive assay for the quantification of tetranor-PGDM in urine.
  • Detect tetranor-PGDM, a major metabolite of PGD2 in urine
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure tetranor-PGDM levels down to 40 pg/ml
  • Read: Colorimetric at 405 to 420 nm
tetranor-PGDM is a major metabolite of prostaglandin D2 (PGD2) found in human and mouse urine. In human urine, tetranor-PGDM is significantly more abundant than the PGD2 metabolites 11β-PGF2α and 2,3-dinor-11β-PGF₂α and is the only endogenous PGD2 metabolite detectable in mouse urine by LC-MS.
Normal levels of tetranor-PGDM in human and mouse urine are 1.5 ng/mg creatinine and 8.1 ng/mg creatinine respectively. tetranor-PGDM ELISA Kit is a competitive assay that can be used for quantification of tetranor-PGDM in urine. This assay converts tetranor-PGDM to a stable derivative, tetranor-PGJM, that can be easily quantified.
Delivery information: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.