Cysteinyl Leukotriene Express ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
10009291-480 10009291-96
75816-728EA 2944.74 CAD
75816-728 75816-730
Cysteinyl Leukotriene Express ELISA Kit, Cayman Chemical Company
Assays ELISAs
Competitive assay used for quantification of CysLT.
  • Detect all CysLTs (LTC4, LTD4, and LTE4) in urine, tissue culture medium, and other sample matrices
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure CysLT levels down to 20 pg/ml
  • Read: Colorimetric at 405 to 420 nm
Leukotrienes (LTs) are a group of acute inflammatory mediators derived from arachidonic acid via the 5-lipoxygenase (5-LO) pathway in leukocytes. LTC4, LTD4, and LTE4 are collectively referred to as cysteinyl leukotrienes (CysLTs).
LTC4 and LTD4 are potent mediators of asthma and hypersensitivity. They induce bronchoconstriction, increase microvascular permeability, and are vasoconstrictors of coronary arteries. The biological activity of LTE4 is much lower in most systems studied, but its presence reflects the prior existence of LTC4 and LTD4.
CysLT Express ELISA Kit is a competitive assay that can be used for measurement of all three primary CysLTs in urine, culture media, and other sample matrices. This sensitive assay takes about three hours to complete.
Delivery information: Kit contains antibody, tracer, buffer a concentrate; wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellmans reagent.