Prostaglandin D₂-MOX Express ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical Company
500151-480 500151-96
75817-258EA 2332.27 CAD
75817-258 75817-260
Prostaglandin D₂-MOX Express ELISA Kit, Cayman Chemical
Assays ELISAs
A competitive assay that permits the rapid measurement of PGD2 from biological samples.
  • Treatment of samples with MOX hydrochloride converts unstable PGD2 to stable PGD2-MOX, MOX derivatization reagent included with assay
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGD2-MOX levels down to 16 pg/ml
  • Rapid assay; get results in under four hours
PGD2-MOX Express ELISA is a competitive assay that permits the rapid measurement of PGD2 from biological samples, requiring only one-hour incubation and development times for each step. The methodology for the assay is the same as that employed in the PGD2-MOX ELISA (Item No. 512011). The ELISA offers the convenience of a fast assay while still achieving a sensitivity of ~16 pg/ml and an assay range of 7.8 to 1000 pg/ml.
Delivery information: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, control, methoxylamine, sodium acetate, tracer and antiserum dye.