Solvent Addition Funnels, Chemglass
Supplier: Chemglass
CG-1758-01 CG-1758-02 CG-1758-03 CG-1758-04 CG-1758-06
80071-192EA 267.9 CAD
80071-192 80071-194 80071-196 80071-198 80071-202
Solvent Addition Funnels, Chemglass
Funnels Dropping Funnels
Funnel design permits addition of solvents to flash chromatography columns without disturbing the silica-gel/packing layer.
  • Lower end of funnel is closed
  • Stem contains a series of holes through which the solvent drains
  • Top of funnel is 75 mm (3")