Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Supplier: Air-Tite
EU032913 EU03308 EU03318 EU052813 EU052913 EU05308 EU05318 EU12558 EU12813 EU12913 EU1308
76290-396BX 37.55 CAD
76290-396 76290-398 76290-400 76290-402 76290-404 76290-406 76290-408 76290-410 76290-412 76290-414 76290-416
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Syringes Syringes, Medical
These EXEL International Insulin Syringes with Fixed Needle are high quality and cost-efficient.
  • Sterile
  • Latex free
  • Non pyrogenic
  • Non toxic
  • Low deadspace
The insulin syringes design includes bold printing, clear graduations and siliconized gaskets for smooth consistent plunger motion and a positive plunger stop. The auto ground needle tip provides for greater comfort. Reduced dead space in this design makes for reduced Insulin waste.
Certifications: Meets ISO Standards, CE registered.
Packaging: Sterile and wrapped packs of 10.
Order Now
SPECIFICATIONS
Environmentally Preferable