Powder Addition Funnels, Chemglass
Supplier: Chemglass
CG-1723-01 CG-1723-08 CG-1723-13 CG-1723-15 CG-1723-16 CG-1723-17 CG-1723-03 CG-1723-05 CG-1723-07 CG-1723-11 CG-1723-12
80070-916EA 898.7 CAD
80070-916 80070-924 80070-932 80070-936 80070-938 80070-940 80070-918 80070-920 80070-922 80070-928 80070-930
Powder Addition Funnels, Chemglass
Funnels Powder Funnels
Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out.
One-piece, easily removed, grooved PTFE auger delivers powders at a uniform rate. Funnel has a lower inner joint and top outer, both being the size listed.
The 250 and 500 ml capacities have the larger, deep cut auger (Size B) for use with coarse material.