Powder Funnels, Chemglass
Supplier: Chemglass
CG-1720-L-01 CG-1720-L-02
80070-866EA 153.62 CAD
80070-866 80070-868
Powder Funnels, Chemglass
Funnels Powder Funnels
Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnel has steep sides to permit free-flow of solid materials. With standard taper inner joint.