Addition Funnels with Threaded Valves, Chemglass
Supplier: Chemglass
CG-1704-04 CG-1704-06 CG-1704-12 CG-1704-01 CG-1704-02 CG-1704-03 CG-1704-10
80070-560EA 855.31 CAD
80070-560 80070-564 80070-572 80070-554 80070-556 80070-558 80070-568
Addition Funnels with Threaded Valves, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm.
  • PTFE plug design permits semi-needle valve action
  • Funnels with 14/20 and 19/22 joints are supplied complete with a stopper
Order Now
SPECIFICATIONS
Environmentally Preferable