One - Hour Fire Barrier, Justrite®
Supplier: Justrite
35352 35354 35356
76292-558EA 2028.73 CAD
76292-558 76292-560 76292-562
One - Hour Fire Barrier, Justrite®
Furniture Storage Cabinets
One-Hour Fire Barrier provides safety when moving and storing of gas cylinders.
  • One hour fire wall rating
  • Each barrier includes six 0.0406" diameter mount holes
Constructed using fire-resistant Marinite® I fully encased into welded steel panels, these units may be mounted to exterior walls and floor using provided mounting holes with self-supplied fasteners, or they may be used as a free-standing wall.
Lift hook plates supplied for installation positioning.