Medical Oxygen Cylinder Holder, Justrite®
Supplier: Justrite
35326 35328 35330
76292-550EA 261.32 CAD
76292-550 76292-552 76292-554
Medical Oxygen Cylinder Holder, Justrite®
Drum Storage
This Medical Oxygen Cylinder Holder provides safety when moving and storing of gas cylinders.
  • Constructed using steel with corrosion-resistant powder coating
  • Stands include 0.312" diameter mounting holes for permanent installations
  • Used for medical oxygen or calibration gases in laboratories
This ring stand uses 10 mm (0.375")nylon set screws to hold cylinders in place.
Certifications: Holders are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA approved.