Gas Cylinder Cart, Justrite®
Supplier: Justrite
35056 35058
76292-518EA 2682.52 CAD
76292-518 76292-520
Gas Cylinder Cart, Justrite®
Trolleys
This Gas Cylinder Cart provides safety when moving and storing of gas cylinders.
  • Carts include two 102 mm (4") locking swivel casters and two fixed direction casters for movement of cylinders with ease
  • Exclusive bumper guard edging with steel reinforced vinyl protects cylinders from damage, while providing extra grip
  • Heavy-duty 38×1372 mm (1.5×54") polypropylene support straps, rated at 516 kg (1138 lbs.)
  • Straps feature steel cinch buckle and hold individual cylinders securely to prevent tipping
Designed for the safe transport of industrial and commercial use compressed gas cylinders, these storage carts store cylinders from 4 to 12" (102 to 305mm) in diameter back-to-back.
Constructed from 7 and 11 gauge (5mm and 3mm) steel, each unit includes upper and lower brackets to provide added support and securement locations for cylinders.
Offset deck design, reinforced with three sets of support bars for strength and rigidity, makes cylinder loading easy and safe at only 1.75" (45mm) off the ground
For an improved level of safety on any installation, mobile stands include openings to retrofit secondary support chains, one for each strap.
Certifications: Carts are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Ordering information: Order model 76292-572 to get optional chain(s).