Double Cylinder Hand Truck with Hoist Ring and Locking Post, Justrite®
Supplier: Justrite
35024 35034 35026 35018 35022
76292-496EA 2631.12 CAD
76292-496 76292-486 76292-488 76292-484 76292-480
Double Cylinder Hand Truck with Hoist Ring and Locking Post, Justrite®
Trolleys
This Double Cylinder Hand Truck with Hoist Ring and Locking Post provides safety when moving and storing of gas cylinders.
  • Durable
  • Provide optimum safety, strength, and longevity
  • Developed for the US Navy for use on submarines
  • Locking posts and collar plates ensure total cylinder immobilization
  • Corrosion-resistant powder-coat finish throughout for smooth handling
  • Floor plate made of thick 11 gauge (3 mm) steel with safety fins prevents warping or breaking
  • Accessorize hand trucks with a tool tray or lockable tool box for securing torches and other equipment
Move gas cylinders easily around a construction site, into locations with poor accessibility, or from floor to floor with these hoist-equipped hand trucks.
Extensive structural framing with 10 mm (0.375") thick hoist plates make these units safe when lifted by hoist rigs. Triple grip handle design helps user control the high center of gravity by expanding available grip area for improved leverage and swing radius.
The zero-lift height loading allows floor plate to lay flat, thereby eliminating the need to lift cylinder during loading and unloading.
Caster carriage model features heavy steel bars and nylon washers for smooth folding action and easy storage in tight spaces.
Two heavy-duty 38×1372 mm (1.5×54") polypropylene support straps, lab tested to 1138 lbs. (516 kg), feature steel cinch buckles for secure attachment of cylinder. Straps eliminate any cylinder rolling potential seen in chain-secured units.
Certifications: Hand Trucks are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.