Single Cryogenic Cylinder Hand Truck, Justrite®
Supplier: Justrite
35010 35012
76292-474EA 1650.55 CAD
76292-474 76292-476
Single Cryogenic Cylinder Hand Truck, Justrite®
Trolleys
This Single Cryogenic Cylinder Hand Truck provides safety when moving and storing of gas cylinders.
  • Rated at 1000 lbs. (454 kg) weight capacity
  • Holds one larger diameter propane or cryogenic cylinder from 9 to 18" (229 to 457 mm)
It has an exclusive bumper guard edging with steel reinforced vinyl, which protects cylinders from damage while providing extra grip.
Available with steel hub pneumatic wheels with dual ball bearings or steel hub, foam filled flat-free wheels with dual ball bearings.