Gas Cylinder Support Bracket, Justrite®
Supplier: Justrite
35250 35252 35254 35256 35258 35260 35262 35264 35266 35268 35270 35272 35274 35276
76292-360EA 86.09 CAD
76292-360 76292-362 76292-364 76292-366 76292-368 76292-370 76292-372 76292-374 76292-376 76292-378 76292-380 76292-382 76292-384 76292-386
Gas Cylinder Support Bracket, Justrite®
Furniture Storage Cabinets
These gas cylinder support brackets provide a quick and easy solution for storing gas cylinders when space is at premium.
  • Brackets accept cylinder sizes ranging from (4 to 12") 102 to 305 mm
  • Exclusive bumper guard edging with steel reinforced vinyl protects cylinders from damage while providing extra grip
  • Designed for wall or bench mounting applications
  • Constructed from heavy gauge, powder-coated steel to ensure long service life and chemical resistance
  • Heavy-duty polypropylene support straps, 1.5×54" (38×1372 mm) and rated at 1138 lbs. (516 kg), feature steel cinch buckle and hold individual cylinders securely against bracket
  • Brackets with chains feature welded and plated, heavy-duty size 2/0 steel chains with twist links allowing links to lay flat against the cylinders, enabling a tighter fit and preventing damage to required labeling
For an improved level of safety for any installation, all brackets include openings to retrofit secondary support chains, one set for each cylinder station.
Certifications: Brackets are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.