Gas Cylinder Forklift Pallets, Individually Secured, Justrite®
Supplier: Justrite
35228 35230 35232 35234
76292-352EA 1346.93 CAD
76292-352 76292-354 76292-356 76292-358
Gas Cylinder Forklift Pallets, Individually Secured, Justrite®
Drum Storage
This Individually Secured Gas Cylinder Forklift Pallet provides safety when moving and storing of gas cylinders.
  • Fully enclosed 197 mm (7.75") wide forklift channels with two-way access
  • Bumper guard edging with steel reinforced vinyl protects cylinders from damage, while providing extra grip
  • Accepts cylinders from 102 to 305 mm (4" to 12") in diameter
  • Secure transportation of multiple gas cylinders with a forklift using these pallets
Fully welded construction from 11 gauge (3 mm) hot-rolled steel, powder-coated to ensure longevity and chemicalMore Product Information