Science Take-Out® Is Climate Change Making Us Sick?
Supplier: SCIENCE TAKE-OUT, LLC
STO-161 STO-161U STO-161R STO-161-NOTG
470311-340EA 25.9 CAD
470311-340 470311-342 470311-354 470311-341
Science Take-Out® Is Climate Change Making Us Sick?
Educational Materials Environmental Educational Materials Environmental Learning Activities
Explore how climate change may lead to health problems.
  • Available as a single kit with material for one student or group
  • Unassembled ten pack for additional savings
  • No additional materials necessary
Jay wonders if climate change might be to blame for increased problems with his allergies. Students conduct simulated antibody tests to determine if Jay is allergic to tree pollen. They graph and analyze data to determine if there is a correlation between temperature, pollen counts, and allergic symptoms.
Caution: Choking hazard. This science take-outMore Product Information