TLG® Cylindrical Separatory Funnels with Teflon® Stopcock, Sati International
Supplier: CHEM SCIENCE INC CA
CS-FC475014 CS-FC471224 CS-FC472540 CS-FC475024
CA10201-652CS 235.79 CAD
CA10201-652 CA10201-654 CA10201-656 CA10201-658
TLG® Cylindrical Separatory Funnels with Teflon® Stopcock, Sati International
Funnels Separating Funnels
Cylindrical shaped separatory funnel has a lower standard taper inner joint and a top outer joint, both being the same size. Graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line.
  • Funnels with 14/20 and 19/22 joints are supplied complete with a stopper
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Annealing point: 560 °C
Linear coefficient of expansion: 32×10-7/°C
Softening point: 820 °C
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”