TLG® Pressure Equalizing Funnels with Double Stopcock, Sati International
Supplier: CHEM SCIENCE INC CA
CS-F0671425G CS-F0671925G CS-F0671460G CS-F0671960G CS-F0672460G CS-F0671125G CS-F0679125G CS-F0674125G CS-F0679250G CS-F0674250G CS-F0674500G CS-F067241LG CS-F0671425 CS-F0671925 CS-F0671460 CS-F0671960 CS-F0672460 CS-F0671125 CS-F0679125 CS-F0674125 CS-F0679250 CS-F0674250 CS-F0674500 CS-F067241L
CA10201-588CS 322.35 CAD
CA10201-588 CA10201-590 CA10201-592 CA10201-594 CA10201-596 CA10201-598 CA10201-600 CA10201-602 CA10201-604 CA10201-606 CA10201-608 CA10201-610 CA10201-612 CA10201-614 CA10201-616 CA10201-618 CA10201-620 CA10201-622 CA10201-624 CA10201-626 CA10203-816 CA10203-818 CA10203-820 CA10203-822
TLG® Pressure Equalizing Funnels with Double Stopcock, Sati International
Funnels Dropping Funnels
Additional straight bore glass or Teflon® stopcock in the pressure equalizing line. Graduation marks on glass.
  • Annealing point: 560 °C
  • Linear coefficient of expansion: 32×10-7/°C
  • Softening point: 820 °C
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”