TLG® Pressure Equalizing Funnels with High Vacuum Valve, Sati International
Supplier: CHEM SCIENCE INC CA
CS-F0641425 CS-F0641460 CS-F0641960 CS-F0642460 CS-F0641125 CS-F0649125 CS-F0644125 CS-F0649250 CS-F0644250
CA10201-628CS 326.13 CAD
CA10201-628 CA10201-630 CA10201-632 CA10201-634 CA10201-636 CA10201-638 CA10201-640 CA10204-272 CA10204-274
TLG® Pressure Equalizing Funnels with High Vacuum Valve, Sati International
Funnels Dropping Funnels
Teflon® extended tip valve for semi-needle valve adjustment.
  • Annealing point: 560 °C
  • Linear coefficient of expansion: 32×10-7/°C
  • Softening point: 820 °C
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”