TLG® Buchner Filter Funnels, Sati International
Supplier: CHEM SCIENCE INC CA
CS-F0601002C CS-F0604060C CS-F0604060M CS-F0604060F CS-F0606150C CS-F0606150M CS-F0601000M CS-F0601000F CS-F0606150F CS-F0608350C CS-F0608350M CS-F0608350F CS-F0609600C CS-F0609600M CS-F0609600F CS-F0601000C CS-F0601002M CS-F0601002F CS-F0602015C CS-F0602015M CS-F0602015F CS-F0603030C CS-F0603030M CS-F0603030F
CA10201-736CS 70.55 CAD
CA10201-736 CA10201-738 CA10201-740 CA10201-742 CA10201-744 CA10201-746 CA10201-748 CA10201-750 CA10203-896 CA10203-898 CA10203-900 CA10203-902 CA10203-904 CA10203-906 CA10203-908 CA10203-910 CA10204-352 CA10204-354 CA10204-356 CA10204-358 CA10204-360 CA10204-362 CA10204-364 CA10204-366
TLG® Buchner Filter Funnels, Sati International
Funnels Filtration Funnels Büchner Funnels
Sealed-in glass disc of the specified porosity. Not recommended for filtration where pressure would exceed 15 psi.
  • Annealing point: 560 °C
  • Linear coefficient of expansion: 32×10-7/°C
  • Softening point: 820 °C
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”