TLG® High Vacuum Valve Funnel Additions, Sati International
Supplier: CHEM SCIENCE INC CA
CS-F0551060 CS-F0554200
CA10201-532CS 471.02 CAD
CA10201-532 CA10201-534
TLG® High Vacuum Valve Funnel Additions, Sati International
Funnels Filtration Funnels Filter Funnels
High vacuum valve contains PTFE O-Ring at tip and PTFE wiper to protect Viton® O-Ring of shaft.
  • Annealing point: 560 °C
  • Linear coefficient of expansion: 32×10-7/°C
  • Softening point: 820 °C
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”