TLG® Separatory Funnels with Teflon® Stopcock, Sati International
Supplier: CHEM SCIENCE INC CA
CS-F0451025 CS-F0451025G CS-F0459025 CS-F0459025G CS-F0451050 CS-F0451050G CS-F0459050 CS-F0459050G CS-F0451125 CS-F0451125A CS-F0459125 CS-F0459125A CS-F0454125 CS-F0454125A CS-F0451250 CS-F0451250A CS-F0459250 CS-F0459250A CS-F0454250 CS-F0454250A
CA10203-846CS 203.44 CAD
CA10203-846 CA10203-848 CA10203-850 CA10203-852 CA10203-854 CA10203-856 CA10203-858 CA10203-860 CA10203-862 CA10204-322 CA10204-324 CA10204-326 CA10204-328 CA10204-330 CA10204-332 CA10204-334 CA10204-336 CA10204-338 CA10204-340 CA10204-342
TLG® Separatory Funnels with Teflon® Stopcock, Sati International
Funnels Separating Funnels
Squibb-type funnel supplied with a Teflon® stopcock. The bottom tip of the funnel has a heavy-duty drip joint. Supplied with a glass or polypropylene stopper.
  • Annealing point: 560 °C
  • Linear coefficient of expansion: 32×10-7/°C
  • Softening point: 820 °C
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”