TLG® High Vacuum Filter Funnels with Fritted Disc, Sati International
Supplier: CHEM SCIENCE INC CA
CS-F0371460C CS-F0371460M CS-F0374200C CS-F0374200M CS-F0401460C CS-F0401460M CS-F0404200C CS-F0404200M
CA10203-792CS 446.51 CAD
CA10203-792 CA10203-794 CA10203-796 CA10203-798 CA10204-242 CA10204-244 CA10204-246 CA10204-248
TLG® High Vacuum Filter Funnels with Fritted Disc, Sati International
Funnels Filtration Funnels Filter Funnels
Funnel features a 0 to 4 mm high vacuum valve and a 2 mm glass stopcock which provides constant pressure retention in the device.
  • Annealing point: 560 °C
  • Linear coefficient of expansion: 32×10-7/°C
  • Softening point: 820 °C
This borosilicate glass with low linear coefficient of expansion is the ideal choice for fabricating laboratory glassware.
Certifications: All TLG® glass products meet ASTM specification E438, Type 1, Class A “Standard Specification for glass in laboratory Apparatus”