Weighing Funnels, Sati International
Supplier: CHEM SCIENCE INC CA
245.202.01 245.202.02 245.202.03
CA10823-034CS 219.88 CAD
CA10823-034 CA10823-036 CA10823-038
Weighing Funnels, Sati International
Boats Weighing Boats
Borosilicate glass funnel with tapered pouring neck for micro transferring of solid and liquid. To be used as weighing dish and sample transfer funnel.
  • Flat bottom to keep funnel stable while weighing