Hirsch Filter Funnels, Sati International
Supplier: CHEM SCIENCE INC CA
239.202.04 239.202.05 239.202.06 239.202.07 239.202.01 239.202.02 239.202.03
CA10201-548CS 151.37 CAD
CA10201-548 CA10201-550 CA10201-552 CA10201-554 CA10203-802 CA10203-804 CA10203-806
Hirsch Filter Funnels, Sati International
Funnels Filtration Funnels Filter Funnels
The conical funnel shape is very useful for preparation work, particularly to concentrate crystal residue for re-washing. The angle of the funnel body facilitates the removal of precipitate.